Learn More
Description
Includes
- Lipoxygenase inhibitor screening assay buffer (10X), 1 vial
- Developing reagent 1, 1 vial
- Developing Reagent 2, 1 vial
- 15-Lipoxygenase standard, 1 vial
- Arachidonic acid (substrate), 1 vial
- Linoleic acid (substrate), 1 vial
- Potassium hydroxide, 1 vial
- 96-Well solid plate (colorimetric Assay), 1 plate
- 96-Well cover sheet, 1 cover
Specifications
Specifications
| Content And Storage | Store the kit at -20°C temporarily. For best results, store the components as described in the protocol. |
| Description | Lipoxygenase Inhibitor Screening Assay Kit is a measurement of detecting hydeoperoxides produced in the lipoxygenation reaction. |
| For Use With (Application) | Functional Study |
| Includes | 1 Vial Lipoxygenase inhibitor screening Assay Buffer 10X, 1 vial developing reagent 1, 1 vial developing reagent 2, 1 vial 15-Lipoxygenase standard, 1 vial Arachidonic Acid (Substrate), 1 vial Linoleic Acid (Substrate), 1 vial Potassium Hydroxide, 1 plate 96-well solid plate (colorimetric assay), 1 cover 96-well cover sheet |
| Quantity | 1 Kit |
| Type | Inhibitor Screening Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.