Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LSD1 Demethylase Activity/Inhibition Assay Kit (Fluorometric)

Catalog No. 89028409 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-409 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-409 Supplier Abnova™ Supplier No. KA1603
Add to Cart
Add to Cart

Used for an easy and fast fluorometric measurement of LSD1 activity or inhibition

Specifications

Content And Storage Store LF3, LF4, LF6 and LF8 at -20°C away from light. Store LF1, LF5, LF7, LF9 and 8-Well Assay Strips at 4°C away from light. Store remaining components (LF2, LF10, Adhesive Covering Film) at room temperature away from light.
Description LSD1 Demethylase Activity/Inhibition Assay Kit (Fluorometric) is used for an easy and fast fluorometric measurement of LSD1 activity or inhibition.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 14mL LF1 (wash buffer 10X), 4mL LF2 (LSD1 assay buffer), 60μL LF3 (LSD1 substrate, 50μg/mL), 10μL LF4 (LSD1 assay standard, 50μg/mL), 5μL LF5 (Capture antibody, 1000
Quantity 1 Kit
Type Activity/inhibition Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.