Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Lysosome/Cytotoxicity Dual Staining Kit is a tool for studying lysosome function at the cellular level
Functional Study; Flow Cytometry
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Immunofluorescence, functional study, flow cytometry and for studying lysosome function at the cellular level |
| Includes | 100μL NBD-PZ stock solution, 5 tablets buffer tablet, 250μL Propidium Iodide solution, 50μL Chloroquine positive control (25mM) |
| Quantity | 1 Kit |
| Type | Dual Staining Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?