Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LYZ (Human) ELISA Kit

Catalog No. 89100237 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, CSF, Plasma, Saliva, Serum, Urine
Milk
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-237 CCS, CSF, Plasma, Saliva, Serum, Urine
89-028-150 Milk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-237 Supplier Abnova™ Supplier No. KA0485
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human LYZ

Specifications

Calibration Range 0.156 to 5ng/mL
Conjugate Biotin
Cross Reactivity Human
Description LYZ (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human LYZ.
Detection Limit 0.15ng/mL
Detection Method Colorimetric
Includes 96 wells-Antibody coated microwells, Human Lysozyme Standard (50ng), Biotinylated Lysozyme Antibody (100X 1vial), 30mL MIXDiluent Concentrate(10x), 2 x 30mL Wash Buffer Concentrate (20X), 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL Chromogen Substrate, 12mL Stop Solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, CSF, Plasma, Saliva, Serum, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type LYZ (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.