Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Magnesium Assay Kit

Catalog No. 89028382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-382 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-382 Supplier Abnova™ Supplier No. KA1650
Add to Cart
Add to Cart

For quantitative colorimetric magnesium determination at 500nm in serum, urine

Specifications

Calibration Range 0.1 to 3mg/dL
Content And Storage Store the kit at 4°C.
Description Magnesium Assay Kit is a quantitative colorimetric magnesium determination at 500 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 25mL Reagent A, 25mL reagent B, 1.5mL x 2 EDTA solution, 1mL standard, 10mg/dL Mg2+
Quantity 1 Kit
Sample Type Serum, Urine
Sample Volume 25 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.