Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Malachite Green Phosphate Assay Kit

Catalog No. 89024615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-615 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-615 Supplier Abnova™ Supplier No. KA1331
Add to Cart
Add to Cart

Includes

  • MG Phosphate Standard (1M) 1 vial/100μL
  • MG Blue Solution 2 vials/1.8mL
  • MG Acidic Solution 2 vials/600μL
  • 96-Well Plate (low volume) 2 plates
  • 96-Well Cover Sheet 2 covers

Used for measuring inorganic free phosphate in aqueous solution, functional study

Specifications

Content And Storage Store the kit at 4°C. For best results, please store the components as described in the protocol.
Description Malachite Green Phosphate Assay Kit is a non-radioactive method for measuring inorganic free phosphate in aqueous solution.
Detection Method Non-radioactive Method
For Use With (Application) Functional Study
Includes 100μL MG Phosphate standard (1M), 1.8mL MG blue solution, 600μL MG Acidic solution, 2 plates 96-well plate (low volume), 2 covers 96-well cover sheet
Quantity 1 Kit
Type Phosphate Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.