Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mbl2 (Mouse) ELISA Kit

Catalog No. 89139333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-333 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-333 Supplier Abnova™ Supplier No. KA4482
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of mouse Mbl2 in cell culture supernates, plasma (EDTA, heparin) and serum samples

  • Sample Type: Cell culture supernates, plasma (heparin, EDTA), serum
  • Sample Volume: 100μL
  • Conjugate: Biotin
  • Reactivity: Mouse
  • Detection Method: Colorimetric
  • Calibration Range: 312 to 20,000 pg/mL
  • Limit of Detection: <10 pg/mL

  • Applications: Quantification
  • Storage: Store the kit at -20°C.

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 312 to 20,000 pg/mL
Description Mbl2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Mbl2.
Detectable Analytes Mbl2
Assay Sandwich ELISA
Conjugate Biotin
Cross Reactivity No detectable cross-reactivity with other relevant proteins
Detection Method Colorimetric
Immunoassay Kit Format 96-well Plate
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant mouse MBL2
Sufficient For 96 Assays
For Use With (Application) Quantification
Gene ID (Entrez) 17195
Host Species Mouse
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at -20°C
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.