Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Methylated DNA Quantification Kit, Fluorometric

Catalog No. 89028206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-206 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-206 Supplier Abnova™ Supplier No. KA1511
Add to Cart
Add to Cart

Detects global DNA methylation status using DNA isolated from any species such as mammals, plants, fungi, bacteria, and viruses in a variety of forms including, but not limited to, cultured cells, fresh and frozen tissues, paraffin-embedded tissues, plasma/serum samples, and body fluid samples.

  • Methylated DNA Quantification Kit (Fluorometric) contains all reagents necessary for the quantification of global DNA methylation
  • This kit is particularly suitable for samples only available in small amounts such as laser capture microdissection samples and embryos

Specifications

Calibration Range 20 to 200ng
Content And Storage Store MF3, MF4, MF6, MF7 and MF8 at -20°C away from light. Store MF1, MF5, MF9 and 8-Well Assay Strips at 4°C away from light. Store MF2 and MF10 at room temperature away from light.
Description Methylated DNA Quantification Kit (Fluorometric) is used for measuring methylated DNA.
Detection Method Fluorometric
For Use With (Application) Quantification
Includes 14mL MF1 (wash buffer 10X), 5mL MF2 (binding solution), 10μL MF3 (negative control, 20μg/mL), 10μL MF4 (positive control, 20μg/mL), 4μL MF5 (Capture antibody, 1000
Quantity 1 Kit
Sample Type DNA Extract
Sample Volume 1 to 8 μL
Type Quantification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.