Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MMP9 (Human) ELISA Kit

Catalog No. 89100793 Shop All Abnova Corporation Products
Catalog No. 89-100-793 Supplier Abnova™ Supplier No. KA3090
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Human MMP-9 antibody, 96 (8 x 12) wells
  • Lyophilized MMP-9 standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Human MMP-9 antibody (Concentrated) Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated) Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively human MMP9.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description MMP9 (Human) ELISA Kit is intended for the quantitative measurement of human MMP9.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Human MMP-9 antibody 96 (8x12) wells, Lyophilized MMP-9 receptor standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Human MMP-9 receptor antibody (Concentrated), Dilution: 1:100 (130mL), Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130UL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <5 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 6 months, or at -20°C for 12 months.
For Use With (Application) Quantification
Product Type MMP9 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.