Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Monoamine Oxidase Assay Kit

Catalog No. 89028364 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-364 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-364 Supplier Abnova™ Supplier No. KA1632
Add to Cart
Add to Cart

For quantitative determination of monoamine oxidase activity in biological sample

Specifications

Content And Storage Store all components at -20°C.
Description Monoamine Oxidase Assay Kit is a quantitative determination of monoamine oxidase activity.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 12mL assay buffer (pH 7.4), 120μL p-Tyramine, 50μL Pargyline (20mM), 120μL HRP enzyme, 50μL Clorgyline (20mM), 120μL dye reagent, 100μL Hydrogen Pe
Quantity 1 Kit
Sample Type Biological Sample
Type Monoamine Oxidase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.