Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MPO Chlorination Assay Kit

Catalog No. 89024618 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-618 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-618 Supplier Abnova™ Supplier No. KA1336
Add to Cart
Add to Cart

Used for detecting the myeloperoxidase chlorination activity and functional study

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description MPO Chlorination Assay Kit is a fluorescence-based method for detecting the myeloperoxidase chlorination activity.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 vial MPO assay buffer, 2 vials MPO chlorination substrate, 1 vial Myeloperoxidase control, 1 vial MPO inhibitor, 1 vial MPO Hydrogen Peroxide, 1 vial Fluorescein standard, 2 plates 96-well plate (black), 2 covers 96-well cover sheets
Quantity 1 Kit
Type Chlorination Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.