Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MPO Peroxidation Assay Kit

Catalog No. 89024620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-620 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-620 Supplier Abnova™ Supplier No. KA1338
Add to Cart
Add to Cart

Fluorescence-based method for detecting the myeloperoxidas peroxidase activity

Functional Study

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description MPO Peroxidation Assay Kit is a fluorescence-based method for detecting the myeloperoxidas peroxidase activity.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 vial MPO assay buffer, 5 vials MPO peroxidation substrate, 1 vial Myeloperoxidase control, 1 vial MPO inhibitor, 1 vial MPO Hydrogen Peroxide, 1 vial MPO Resorufin standard, 1 vial MPO DMSO, 2 plates 96-well plate (black), 2 covers 96-well cover sheets
Quantity 1 Kit
Type Peroxidation Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.