Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Multi-Parameter Apoptosis Assay Kit

Catalog No. 89024617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-617 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-617 Supplier Abnova™ Supplier No. KA1335

missing translation for 'validMainframeMsg'

Multi-Parameter Apoptosis Assay Kit is a measurement for detecting apoptosis cells.

Immunofluorescence, Functional Study, Flow Cytometry

Specifications

Quantity 1 Kit
Content And Storage -20°C
For Use With (Application) Apoptosis Detection, Flow Cytometry, Functional Study, Immunofluorescence
Conjugate FITC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.