Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NAD+/NADH Assay Kit

Catalog No. 89028389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-389 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-389 Supplier Abnova™ Supplier No. KA1657
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ultrasensitive colorimetric determination of NAD+/NADH at 565nm in cell, tissue

Specifications

Calibration Range 0.05 to 10μM
Content And Storage Store the kit at -20°C.
Description NAD+/NADH Assay Kit is ultrasensitive colorimetric determination of NAD+/NADH at 565 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 10mL assay buffer, 1.5mL Lactate, 1.5mL MTT solution, 12mL NAD/NADH Extraction buffer, 120μL enzyme A, 120μL enzyme B, 0.5mL NAD standard
Quantity 1 Kit
Sample Type Cell, Tissue
Sample Volume 40 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.