Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NADP+/NADPH Assay Kit

Catalog No. 89028395 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-395 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-395 Supplier Abnova™ Supplier No. KA1663
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Assay buffer, 10mL
  • Glucose (1 M), 1.5mL
  • MTT solution, 1.5mL
  • Enzyme mix, 120μL
  • NADP standard, 0.5mL (1mM)
  • NADP/NADPH extraction buffers each, 12mL

Ultrasensitive colorimetric determination of NADP+/NADPH at 565nm

Specifications

Calibration Range 0.1 to 10μM
Content And Storage Store the kit at -20°C.
Description NADP+/NADPH Assay Kit is a ultrasensitive colorimetric determination of NADP+/NADPH at 565 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 10mL assay buffer, 1.5mL Glucose (1M), 1.5mL MTT solution, 120μL enzyme mix, 0.5mL 1mM NADP standard, 12mL NADP/NADPH Extraction buffers
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.