Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Neuraminidase Assay Kit

Catalog No. 89028365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-365 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-365 Supplier Abnova™ Supplier No. KA1633
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Assay buffer, 6mL
  • Substrate, 6mL
  • Cofactors, 120μL
  • Dye reagent, 60μL
  • Enzyme, 120μL
  • Standard, 500μL

For quantitative colorimetric/fluorometer determination of neuraminidase activity in biological sample

Specifications

Calibration Range 0.1 to 10U/L colorimetric assay, 0.01 to 2U/L fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Neuraminidase Assay Kit is a quantitative colorimetric/fluorimetric determination of neuraminidase activity.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 6mL assay buffer, 6mL substrate, 120μL Co-factor, 60μL dye reagent, 120μL enzyme, 500μL standard
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 20 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.