Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Nitrate/Nitrite Fluorometric Assay Kit

Catalog No. 89024626 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-626 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-626 Supplier Abnova™ Supplier No. KA1344
Add to Cart
Add to Cart

Used for detecting total nitrate/nitrite concentration and functional study

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description Nitrate/Nitrite Fluorometric Assay Kit is a measurement for detecting total nitratr/nitrite concentration.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 Vial Nitrate/Nitrite Assay Buffer, 2 vials Nitrate Reductase Enzyme preparation, 2 vials Nitrate Reductase Co-factor preparation, 1 vial Nitrate standard, 1 vial Nitrite standard, 1 vial DAN reagent, 1 vial Sodium Hydroxide (2.8M),3 plates 96-well plate (white), 3 covers 96-well cover sheet
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.