Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Nitric Oxide Assay Kit

Catalog No. 89028373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-373 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-373 Supplier Abnova™ Supplier No. KA1641
Add to Cart
Add to Cart

For quantitative colorimetric determination of nitric oxide at 540nm in cell, food sample, plasma, serum, tissue, urine

Specifications

Content And Storage Store standard at -20°C. Store all other components at -20 to 4°C.
Description Nitric Oxide Assay Kit is a quantitative colorimetric determination of Nitric Oxide at 540 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 12mL Reagent A, 500μL reagent B, 12mL reagent C, 1mL NaOH, 1mL ZnSO4, 1mL standard
Quantity 1 Kit
Sample Type Cell, Food Sample, Plasma, Serum, Tissue, Urine
Sample Volume 100 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.