Learn More
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | 10mL assay buffer, 600μL substrate, 120μL GDH, 12mL reagent A, 500μL reagent B, 12mL reagent C, 600μL reagent D, 1.5mL reagent E, 1mL ZnSO4, 1mL standard, 1mL NaOH. Store Assay Buffer, Substrate, Reagent D, Reagent E and GDH at -20°C. Store other reagents at 4°C. |
| For Use With (Application) | Functional Study |
| Concentration | 0.25 to 25U/L |
| Detection Method | Colorimetric |
| Excitation/Emission | 540 nm |
| Product Type | Detection Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.