Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NOS Activity Assay Kit

Catalog No. 89024627 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-627 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-627 Supplier Abnova™ Supplier No. KA1345
Add to Cart
Add to Cart

Used for detecting nitric oxide synthase activity and functional study

Specifications

Content And Storage Store the kit at -80°C temporarily. For best results, store the components as described in the protocol.
Description NOS Activity Assay Kit is a method for detecting nitric oxide synthase activity.
For Use With (Application) Functional Study
Includes 50μL iNOS positive control, 200μL Calmodulin (1μM), 1.25mL reaction buffer 2X, 100μL L-NNA (10mM), 50mL homogenization buffer 10X, 25mL Stop buffer, 5mL equilibrated Resin, 400
Quantity 1 Kit
Type Activity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.