Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Nuclear Extraction Kit

Catalog No. 89024628 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-628 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-628 Supplier Abnova™ Supplier No. KA1346
Add to Cart
Add to Cart

Includes

  • Nuclear extraction hypotonic buffer (10X), 1 vial/6mL
  • Nuclear extraction dithiothreitol (1 M), 1 vial/60μL
  • Nuclear extraction protease inhibitor cocktail (100X), 1 vial/650μL
  • Nuclear extraction PBS (10X), 1 vial/100mL
  • Nuclear extraction phosphatase inhibitors (50X), 5 vials/5mL
  • Nuclear extraction buffer (2X), 1 vial/3mL
  • Nonidet P-40 assay reagent (10%), 1 vial/6mL

Used for preparing nuclear extraction and functional study

Specifications

Content And Storage Store the inhibitors at -20°C.
Description Nuclear Extraction Kit is a method for prepareing nuclear extracion.
For Use With (Application) Functional Study
Includes 6mL Nuclear Extraction Hypotonic buffer 10X, 60μL nuclear extraction Dithiothreitol (1M), 650μL nuclear extraction Protease inhibitor cocktail 100X, 100mL nuclear extraction PBS 10X, 5mL nuclear extraction Phosphatase i
Quantity 1 Kit
Type Nuclear Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.