Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ORM1 (Rat) ELISA Kit

Catalog No. 89100402 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Biological Fluid
Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-402 Biological Fluid
89-100-680 Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-402 Supplier Abnova™ Supplier No. KA2133
Add to Cart
Add to Cart

Includes

  • Diluent Concentrate (Running Buffer)
  • Wash Solution Concentrate
  • Enzyme-Antibody Conjugate 100X: affinity-purified anti-Rat Alpha 1-Acid Glycoprotein antibody conjugated with horseradish peroxidase in a stabilizing buffer
  • Chromogen-Substrate Solution
  • Stop Solution: 0.3M sulfuric acid. WARNING: Avoid contact with skin
  • Anti- Rat Alpha 1-Acid Glycoprotein ELISA Micro Plate: Twelve removable eight (8) well micro well strips in well holder frame. Each well is coated with affinity-purified anti-Rat AGP
  • Rat Alpha 1-Acid Glycoprotein Calibrator: a lyophilized Rat Alpha 1-Acid Glycoprotein calibrator

Orm1 (Rat) ELISA Kit is based on a solid phase enzyme-linked immunosorbent assay (ELISA)

Specifications

Calibration Range 12.5 to 400ng/mL
Conjugate HRP
Cross Reactivity Rat
Description Orm1 (Rat) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of rat Orm1.
Detection Method Colorimetric
Includes Diluent Concentrate (50mL), Wash Solution Concentrate (50mL), Enzyme-Antibody Conjugate Concentrate (150μL), TMB Substrate Solution (12mL), Stop solution (12mL), Microtiter Plate (96 well), 1 vial Calibrator (Lyophilized)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological Fluid
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.