Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Oxaloacetate Assay Kit

Catalog No. 89101208 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
Colorimetric: 7 to 400μM, Fluorometric: 1 to 40μM
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range
89-101-208 Colorimetric: 7 to 400μM, Fluorometric: 1 to 40μM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-208 Supplier Abnova™ Supplier No. KA3793
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for quantifying OAA and functional study

Specifications

Calibration Range Colorimetric: 7 to 400μM, Fluorometric: 1 to 40μM
Content And Storage Store the kit at -20°C.
Description Oxaloacetate Assay Kit is a quantitative colorimetric/fluorimetric determination of oxaloacetate at OD570, FL530/585 nm.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Quantity 1 Kit
Type Oxaloacetate Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.