Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PAF Acetylhydrolase Assay Kit

Catalog No. 89024636 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-636 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-636 Supplier Abnova™ Supplier No. KA1354
Add to Cart
Add to Cart

Used for measurement of platelet-activating factor acetylhydrolase activity (both cytosolic and extracellular) and functional study

Specifications

Content And Storage Store the kit at -20°C.
Description PAF Acetylhydrolase Assay Kit provides an accurate and method for measurement of platelet-activating factor acetylhydrolase activity (both cytosolic and extracellular).
For Use With (Application) Functional Study
Includes 1 vial PAF Acetylhydrolase assay buffer 1, 4 vials DTNB, 2 vials 2-thio PAF (substrate), 1 vial human Plasma PAF-AH standard, 1 vial PAF Acetylhydrolase assay buffer 2, 1 plate 96-well solid plate (colorimetric assay), 1 cover 96-well cover sheet
Quantity 1 Kit
Type PAF Acetylhydrolase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.