Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PAF Acetylhydrolase Inhibitor Screening Assay Kit

Catalog No. 89024637 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-637 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-637 Supplier Abnova™ Supplier No. KA1355
Add to Cart
Add to Cart

Used for screening vast numbers of inhibitors of platelet-activating factor acetylhydrolase and functional study

Specifications

Content And Storage Store the kit at -20°C.
Description PAF Acetylhydrolase Inhibitor Screening Assay Kit is used for screening vast numbers of inhibitors of platelet-activating factor acetylhydrolase.
For Use With (Application) Functional Study
Includes 1 vial assay buffer 10X, 4 vials DTNB, 2 vials 2-thio PAF (substrate), 2 vials human plasma PAF-AH, 1 plate 96-well plate, 1 cover 96-well cover sheet
Quantity 1 Kit
Type Inhibitor Screening Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.