Learn More
Description
Peroxidase Assay Kit is a quantitative colorimetric/fluorimetric peroxidase determinations
Functional Study
Specifications
Specifications
| Calibration Range | 4 to 1000IU/L |
| Content And Storage | Assay buffer at 4°C and other reagents at -20°C |
| Detection Method | Colorimetric, Fluorometric |
| For Use With (Application) | Quantitative colorimetric/fluorometer peroxidase determinations in biological sample, plasma, saliva, serum, urine |
| Includes | 20mL assay buffer (pH 7), 100μL 3% stabilized H2O2, 5mL (equivalent to 1000 IU/L) calibrator, 60μL dye reagent, 12mL stop reagent |
| Quantity | 100 Assays |
| Sample Type | Biological Sample, Plasma, Saliva, Serum, Urine |
| Sample Volume | 10 μL |
| Type | Peroxidase Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.