Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Peroxide Assay Kit

Catalog No. 89028350 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-350 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-350 Supplier Abnova™ Supplier No. KA1618
Add to Cart
Add to Cart

For quantitative colorimetric peroxide determination at 585nm in cell lysate, culture medium, plasma (citrate), serum, urine

Specifications

Calibration Range 0.2 to 30μM
Content And Storage Store the kit at 4°C.
Description Peroxide Assay Kit is a quantitative colorimetric peroxide determination at 585 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 1mL Reagent A, 50mL reagent B, 100μL standard 3% stabilized H2O2
Quantity 1 Kit
Sample Type Cell Lysate, Culture Medium, Plasma (Citrate), Serum, Urine
Sample Volume 40 μL
Type Peroxide Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.