Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phospholipase D Assay Kit

Catalog No. 89028368 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-368 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-368 Supplier Abnova™ Supplier No. KA1636
Add to Cart
Add to Cart

Includes

  • Assay buffer, 10mL
  • Dye reagent, 120μL
  • Enzyme mix, 120μL
  • Substrate, 1.5mL
  • Calibrator, 400μL

For quantitative colorimetric/fluorometer phospholipase D activity determination in biological sample

Specifications

Calibration Range 0.06 to 10U/L colorimetric assay, 0.04 to 1U/L fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Phospholipase D Assay Kit is a quantitative colorimetric/fluorimetric phospholipase D activity determination.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 10mL assay buffer, 120μL dye reagent, enzyme mix (Dried) 1 vial, 1.5mL substrate, 400μL calibrator
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 10 μL
Type Phospholipase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.