Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phospholipases A2 (calcium-dependent cytosolic) Assay Kit

Catalog No. 89024597 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-597 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-597 Supplier Abnova™ Supplier No. KA1307
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for assaying the activity of cPLA2 and functional study

Specifications

Content And Storage Store the kit at -20°C.
Description Phospholipases A2 (calcium-dependent cytosolic) Assay Kit is a colorimetric method for assaying the activity of cPLA2.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 1 vial cPLA2 assay buffer, 4 vials DTNB/EGTA, 2 vials arachidonoyl Thio-PC (substrate), 1 vial bee venom PLA2 control, 1 vial bromoenol lactone solution, 96-well solid plate (colorimetric assay), 1 cover 96 well cover sheet
Quantity 1 Kit
Type Phospholipase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.