Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phospholipid Assay Kit

Catalog No. 89028367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-367 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-367 Supplier Abnova™ Supplier No. KA1635
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Assay buffer, 10mL
  • PLD Enzyme, 120μL
  • Enzyme mix, 120μL
  • Dye reagent, 120μL
  • Standard, 400μL (2mM phosphatidylcholine)

For quantitative colorimetric/fluorometer phospholipid determination in plasma (no EDTA), serum

Specifications

Calibration Range 3 to 200μM colorimetric assay, 0.6 to 20μM fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Phospholipid Assay Kit is a quantitative colorimetric/fluorimetric phospholipid determination.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 10mL assay buffer, 120μL PLD enzyme, enzyme mix (Dried) 1 vial, 120μL dye reagent, 400μL standard Phosphatidylcholine (2mM)
Quantity 1 Kit
Sample Type Plasma (no EDTA), Serum
Sample Volume 20 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.