Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PLAU (Human) Chromogenic Activity Assay Kit (Direct Assay)

Catalog No. 89028043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-043 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-043 Supplier Abnova™ Supplier No. KA0976
Add to Cart
Add to Cart

Used to determine Human PLAU activity in plasma and cell culture supernatants and functional study

Specifications

Content And Storage Store the kit at 4°C.
Description PLAU (Human) Chromogenic Activity Assay Kit (Direct Assay) is developed to determine human PLAU activity in plasma and cell culture supernatants.
For Use With (Application) Functional Study
Includes 96-well Polystyrene microplate, sealing tapes, 30mL assay diluent, 1 vial human high molecular weight uPA (1200 IU), 2 tubes uPA substrate (lyophilized)
Quantity 1 Kit
Type Chromogenic Activity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.