Learn More
Abnova™ PLAU (Human) Chromogenic Activity Assay Kit (Indirect Assay)
Catalog No. 89028042 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-028-042 1 Kit
1 options
Catalog No. 89-028-042 Supplier Abnova™ Supplier No. KA0977
Add to Cart
Add to Cart
Used to determine Human PLAU activity in plasma and cell culture supernatants and functional study
Specifications
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
Description PLAU (Human) Chromogenic Activity Assay Kit (Indirect Assay) is developed to determine human PLAU activity in plasma and cell culture supernatants.
For Use With (Application) Functional Study
Includes 96-well Polystyrene microplate, sealing tapes, 30mL assay diluent, 1 vial human high molecular weight uPA (1200 IU), 1 vial human Plasminogen, 2 vials Plasmin substrate
Quantity 1 Kit
Type Chromogenic Activity Assay Kit