Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ pNPP Phosphatase Assay Kit

Catalog No. 89028369 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-369 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-369 Supplier Abnova™ Supplier No. KA1637
Add to Cart
Add to Cart

Colorimetric determination of phosphatase activity at 405nm

Specifications

Content And Storage Store Reagent at -20°C. Store other component at 4°C.
Description pNPP Phosphatase Assay Kit is a colorimetric determination of phosphatase activity at 405 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 280μL Reagent, 25mL assay buffer, 25mL stop solution
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.