Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Prl (Mouse/Rat) ELISA Kit

Catalog No. 89024464 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-464 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-464 Supplier Abnova™ Supplier No. KA0923
Add to Cart
Add to Cart

Includes

  • Microwell coated with Prolactin polyclonal antibody, 12 x 8 x 1 96 Tests
  • Prolactin Standards: 6 vials (Ready to use), 0.5mL
  • Biotinylated Antibody reagent, 12mL
  • Streptavidin Enzyme Conjugate: 1 bottle (Ready to use), 12mL
  • TMB Substrate: 1 bottle (Ready to use), 12mL
  • Stop Solution: 1 bottle (Ready to use), 12mL
  • 20X Wash concentrate: 1 bottle, 25mL

For the quantitative measurement of Prolactin in a Mouse/rat Plasma

Specifications

Calibration Range 0.2 to 200ng/mL
Conjugate Biotin
Cross Reactivity Mouse, Rat
Description Prl (Mouse/Rat) ELISA Kit is used for the quantitative measurement of prolactin in a mouse/rat plasma.
Detection Method Colorimetric
Includes Microwell coated with Prolactin Polyclonal Antibody 12 x 8 x 1, 0.5mL Prolactin standards, 12mL Biotinylated Antibody reagent, 12mL Streptavidin Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution, 25mL (20X) Wash concentrate
Quantity 96 Assays
Sample Type Plasma
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Prl (Mouse/Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.