Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Procalcitonin (Human) ELISA Kit

Catalog No. 89139157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-157 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-157 Supplier Abnova™ Supplier No. KA1763

In vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Procalcitonin in cell culture supernatant, plasma, serum and urine samples

  • Sample Type: Cell culture supernatant, plasma, serum and urine
  • Sample Volume: 100μL
  • Conjugate: Biotin
  • Reactivity: Human
  • Detection Method: Colorimetric
  • Limit of Detection: <30 pg/mL

  • Applications: Quantification
  • Storage: Store the kit at -4°C.

Specifications

Assay Sensitivity <30 pg/mL
Description Procalcitonin (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Procalcitonin.
Detectable Analytes Procalcitonin
Assay In vitro ELISA
Conjugate Biotin
Detection Method Colorimetric
Immunoassay Kit Format 96-well Plate
Product Type Procalcitonin (Human) ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Specificity No cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1a, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-γ, L
Sufficient For 96 Assays
Target Species Human
For Use With (Application) Quantification
Host Species Human
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.