Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Prostaglandin F2 alpha ELISA Kit

Catalog No. 89100176 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Milk, Plasma, Saliva, Serum, Urine
CCS, Plasma, Saliva, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-176 CCS, Milk, Plasma, Saliva, Serum, Urine
89-100-161 CCS, Plasma, Saliva, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-176 Supplier Abnova™ Supplier No. KA0327
Add to Cart
Add to Cart

Includes

  • Donkey anti-Sheep IgG Microtiter Plate, 96 Wells
  • PGF2α HS ELISA Conjugate, 2.5mL
  • PGF2α HS ELISA Antibody, 2.5mL
  • Assay Buffer, 30mL
  • Wash Buffer Concentrate, 27mL
  • Prostaglandin F2α Standard, 0.5mL
  • pNpp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 1 slide

The Prostaglandin F2 alpha ELISA Kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2α in milk, culture supernates, saliva, urine, serum and plasma

Specifications

Calibration Range 1.95 to 2000pg/mL
Conjugate AP
Description PGF2-alpha High Sensitivity ELISA Kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2-alpha in milk, culture supernates, saliva, urine, serum and plasma.
Detection Limit 0.98pg/mL
Detection Method Colorimetric
Includes Microwells (96 wells), EIA Conjugate (5mL), EIA Antibody (5mL), Assay Buffer (30mL), Wash Buffer Concentrate (30mL), Standard (0.5mL), pNpp Substrate (20mL), Stop Solution: (5mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 100 μL
Sensitivity 0.98 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.