Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Protein Assay Kit (Fluorometric)

Catalog No. 89101199 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-199 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-199 Supplier Abnova™ Supplier No. KA3783
Add to Cart
Add to Cart

Quantitative fluorimetric determination of protein at FL360/450nm.

Specifications

Calibration Range 0.05 to 200μg/mL
Content And Storage Store the kit at -20°C.
Description Protein Assay Kit is a quantitative fluorimetric determination of protein at FL360/450 nm.
Detection Method Fluorometric
For Use With (Application) Functional Study
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 10 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.