Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Protein Assay Kit

Catalog No. 89028370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-370 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-370 Supplier Abnova™ Supplier No. KA1638
Add to Cart
Add to Cart

Protein Assay Kit is a bradford colorimetric protein determination at 595 nm.

  • Sensitive and accurate: Use 10 μL samples
  • Detection range 0.06 – 1.0 mg /mL protein in 96-well plate assay
  • Simple and high-throughput: the “mix-and-read” procedure involves addition of a single working reagent and reading the optical density; can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day
  • Low interference: Glucose, Tris, vitamins, and amino acids, DNA, RNA, salts, EDTA, phenol, urea, Triton, and SDS do not interfere in the assay
  • Versatility: assays can be executed in 96-well plate or cuvette
  • Store the reagent at 4°C and standard at -20°C, respectively; shelf life of 12 months after receipt
  • Reagents are for research use only

Specifications

Description Protein Assay Kit is a bradford colorimetric protein determination at 595 nm.
For Use With (Application) Functional Study
Includes 20mL Reagent, 1mL standard
Sample Volume 10μL
Content And Storage Store Reagent at 4°C. Store Standard at -20°C.
Product Type Protein Assay Kit
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.