Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Protein Expression Kit II (GST-Fusion Protein)

Catalog No. 89028191 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-191 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-191 Supplier Abnova™ Supplier No. KA1438
Add to Cart
Add to Cart
Request a Quote

This kit produces GST-fusion proteins at high purities and high yields. The wheat germ included in this kit is a specialized high yield wheat germ extract prepared by removing endogenous proteins as nonspecifically adsorb to GST-tag affinity purification resin.

  • WEPRO7240 Expression Kit contains WEPRO™ 7240, suitable for protein expression where affinity purification to high purities is neither necessary nor desirable
  • WEPRO7240H Expression Kit is suitable for His-tagged protein expression and purification; the post-purification purity of the protein expressed with this kit is higher than that with WEPRO7240 Expression Kit
  • WEPRO7240G Expression Kit is suitable for GST-tagged protein expression and purification; the post-purification purity of the protein expressed with this kit is higher than that with WEPRO7240 Expression Kit
  • For research use only

Specifications

Description This kit produces GST-fusion proteins at high purities and high yields. The wheat germ included in this kit is a specialized high yield wheat germ extract prepared by removing endogenous proteins as nonspecifically adsorb to GST-tag affinity purification resin.
For Use With (Application) Produces GST fusion Proteins at high purities and high yields
Content And Storage Store the component at -20°C or -80°C as shown on the protocol.
Product Type Protein Expression Kit II (GST-Fusion Protein)
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.