Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Pyruvate Assay Kit

Catalog No. 89028406 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-406 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-406 Supplier Abnova™ Supplier No. KA1674
Add to Cart
Add to Cart

For quantitative colorimetric/fluorometer pyruvate determination in biological sample

Specifications

Calibration Range 2 to 500μM colorimetric assay, 0.2 to 50μM fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Pyruvate Assay Kit is a quantitative colorimetric/fluorimetric pyruvate determination.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 10mL enzyme mix, 120μL dye reagent, 400μL 25mM Pyruvate standard
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 10 μL
Type Pyruvate Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.