Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Pyruvate Kinase Assay Kit

Catalog No. 89101209 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Cell, Plasma, Serum, Tissue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-209 Cell, Plasma, Serum, Tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-209 Supplier Abnova™ Supplier No. KA3794
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for measuring pyruvate kinase activity and functional study

Specifications

Calibration Range Colorimetric: 0.1 to 50μ/L, Fluorometric: 0.01 to 2μ/L
Content And Storage Store the kit at -20°C.
Description Pyruvate Kinase Assay Kit is a quantitative colorimetric/fluorimetric determination of pyruvate kinase at OD570, FL530/585 nm.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Quantity 1 Kit
Sample Type Cell, Plasma, Serum, Tissue
Type Kinase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.