Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RBP4 (Human) ELISA Kit

Catalog No. 89028124 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Volume:
50 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
89-028-124 0.1 mL
89-100-224 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-124 Supplier Abnova™ Supplier No. KA0989
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human RBP4

Specifications

Calibration Range 312 to 20,000 pg/mL
Content And Storage Store the kit at -20°C
Description RBP4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human RBP4.
Conjugate Biotin
Cross Reactivity Human
Detection Method Colorimetric
Product Type RBP4 (Human) ELISA kit
Sample Type Body Fluids, Cell Culture Supernatant, Plasma, Serum, Tissue Lysates
Sufficient For 96 Tests
For Use With (Application) Quantification
Kit Contents 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Human RBP4 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human RBP4 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin -Biotin
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 0.1 mL
Detection Limit 10 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.