Learn More
Specifications
Specifications
| Calibration Range | 6.25 to 400 ng/mL |
| Content And Storage | Store components of the kit at 4°C or -20°C as described in the protocol. |
| Description | RBP4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human RBP4. |
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Product Type | RBP4 (Human) ELISA kit |
| Sample Type | Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine |
| Sufficient For | 96 Tests |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.