Learn More
Specifications
Specifications
| Content And Storage | Store the kit at 4°C for 6 months. For long term storage, please store at -20°C for 12 months. |
| Description | Rheumatoid factor IgG ELISA Kit is a sandwich enzyme immunoassay for for the quantitative measurement of IgG, IgM and IgA rheumatoid factors in human serum or plasma. |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Product Type | Rheumatoid factor IgG ELISA kit |
| Sample Type | Serum and Plasma |
| For Use With (Application) | Quantification |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Volume | 100 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.