Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SAA (Human) ELISA Kit

Catalog No. 89022945 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-945 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-945 Supplier Abnova™ Supplier No. KA0518
Add to Cart
Add to Cart

Includes

  • Microtiter plate, Pre-coated with anti-human SAA monoclonal antibody, 96 wells
  • Conjugate, Anti-human SAA polyclonal antibody conjugated to horseradish peroxidase with preservative, 15mL
  • Standard, Recombinant human SAA (160ng/vial) in a buffered protein base with preservative, lyophilized, 2 vial
  • Calibrator diluent I, Animal serum with preservative. For serum/plasma testing, 30mL
  • Calibrator diluent II Cell culture medium with calf serum and preservative. For cell culture supernatant testing, 30mL
  • Wash Buffer (20X), 20-fold concentrated solution of buffered surfactant, 60mL
  • Substrate A, Buffered solution with H2O2, 11mL
  • Substrate B, Buffered solution with TMB, 11mL
  • Stop solution, 2N Sulphuric Acid (H2SO4) (Caution: Caustic Material), 14mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human SAA

Specifications

Calibration Range 1.1 to 80ng/mL
Conjugate HRP
Cross Reactivity Human
Description SAA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SAA.
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 15mL SAA Conjugate, 160ng/vial x 2 SAA standard, 30mL calibrator Diluent I, 30mL calibrator Diluent II, 60mL Wash Buffer 20X, 11mL Substrate A, 11mL Substrate B, 14mL stop solution
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type SAA (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.