Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Sialic Acid Assay Kit

Catalog No. 89028387 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-387 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-387 Supplier Abnova™ Supplier No. KA1655
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Dye reagent, 6mL
  • 10% TCA, 5mL
  • DMSO, 12mL
  • Oxidation reagent, 10mL
  • Hydrolysis reagent, 10mL
  • Standard: 10mM sialic acid, 500μL

For quantitative determination of free and total sialic acid in milk, plasma, saliva, serum

Specifications

Calibration Range 5 to 1000μM colorimetric assay, 0.5 to 100μM fluorimetric assay
Content And Storage Store the standard at -20°C. Store all other components at room temperature.
Description Sialic Acid Assay Kit is a quantitative determination of free and total sialic acid.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes 6mL dye reagent, 5mL 10% TCA, 12mL DMSO, 10mL Oxidation reagent, 10mL hydrolysis reagent, 500μL standard 10mM Sialic Acid
Quantity 1 Kit
Sample Type Milk, Plasma, Saliva, Serum
Sample Volume 60 μL
Type Sialic Acid Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.