Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SIRT1 FRET-Based Screening Assay Kit

Catalog No. 89024645 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-645 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-645 Supplier Abnova™ Supplier No. KA1367
Add to Cart
Add to Cart

Used for screening SIRT1 inhibitors or activators and functional study

Specifications

Content And Storage Store the kit at -80°C.
Description SIRT1 FRET-based Screening Assay Kit provides a fluorescence-based method for screening SIRT1 inhibitors or activators.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 Vial SIRT1 FRET Assay Buffer 10X, 1 vial SIRT1 FRET (Human recombinant), 2 vials SIRT1 FRET Peptide, 1 vial SIRT1 FRET NAD+, 1 vial SIRT1 FRET stop solution, 1 vial SIRT1 FRET developer, 1 vial SIRT1 FRET Fluorophore, 1 plate half volume 96-well plate (white), 1 cover 96-well cover sheet
Quantity 1 Kit
Type FRET-Based Screening Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.