Learn More
Specifications
Specifications
| Content And Storage | Store the kit at -80°C temporarily. For best results, store the components as described in the protocol. |
| Description | SIRT2 Direct Fluorescent Screening Assay provides a fluorescence-based method for screening SIRT2 inhibitors or activators. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 1 vial SIRT2 direct assay buffer 10X, 2 vials SIRT2 (human recombinant), 2 vials SIRT2 direct Peptide, 1 vial SIRT2 Direct NAD+, 1 vial SIRT2 direct Nicotinamide, 1 vial SIRT2 direct developer, 1 vial SIRT2 direct Fluorophore, 1 plate half-volume 96-well |
| Quantity | 1 Kit |
| Type | Fluorescent Screening Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.