Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SIRT3 Direct Fluorescent Screening Assay Kit

Catalog No. 89024647 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-647 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-647 Supplier Abnova™ Supplier No. KA1369
Add to Cart
Add to Cart

Used for screening SIRT3 inhibitors or activators and functional study

Specifications

Content And Storage Store the kit at -80°C.
Description SIRT3 Direct Fluorescent Screening Assay Kit provides a fluorescence-based method for screening SIRT3 inhibitors or activators.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 vial assay buffer 10X, 2 vials SIRT3 (human recombinant), 2 vials SIRT3 direct peptide, 1 vial NAD+, 1 vial nicotinamide, 1 vial developer, 1 vial fluorophore, 1 plate half volume 96-well plate (white), 1 cover 96-well cover sheet
Quantity 1 Kit
Type Fluorescent Screening Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.