Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ sPLA2 activity Kit

Catalog No. 89028175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-175 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-175 Supplier Abnova™ Supplier No. KA1015
Add to Cart
Add to Cart

Used for the quantitative measurement of secretory phospholipase A2's (sPLA2) in culture supernatants, cell lysates and buffers from any species

Specifications

Calibration Range 5 to 80U/mL
Content And Storage Store substrate at -20°C. Store all other components at 4°C.
Description sPLA2 activity Kit is a higher throughput assay for the quantitative measurement of sPLA2 in culture supernatants, cell lysates, and buffers from any species.
Detection Method Colorimetric
For Use With (Application) Quantification
Includes 1 each microtiter plate, 2 vials sPLA2 standard, 15mL sPLA2 reaction buffer concentrate, 2 vials sPLA2 substrate, 3mL stop reagent, 3mL sPLA2 color reagent, 1 each plate sealer
Quantity 1 Kit
Sample Type Cell Lysate, Culture Supernatant
Sample Volume 50 μL
Type Activity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.